Invitrogen
Manufacturer Code:MA535587
Catalog # PIMA535587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0255 |
Immunogen | A synthesized peptide derived from human Histone H3.3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: BOS_15646 CG5825 CG8989 dH3.3A DmelCG5825 Dmel_CG5825 H3.3 H3.3a H3.3B H3.4 H3.A H3.B H3/g H3-3a H3-3b H3B H3F3 H3F3A H3f3b H3FT H3t His3.3 His3.3A His-3.3A His3.3B Histone cluster 3 Histone H3 Histone H3.3 Histone H3.3A Histone H3.3B MGC126886 MGC126888 PH3 PP781; Histone H3.3
Gene Aliases: H3-3A; H3-3B; H3.3A; H3.3B; H3F3; H3F3A; H3F3B; PP781
UniProt ID: (Human) P84243, (Mouse) P84244
Entrez Gene ID: (Human) 3020, (Mouse) 15078
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.