Invitrogen
Manufacturer Code:PA523288
Catalog # PIPA523288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (119 TIMPKDIQLARRIRGERA 136) of Histone H3.3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG5825-PA; CG5825-PC; CG5825-PD; family 3B (H3.3B); H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3.3A/B; H3.A; H3.B; H3L-like histone; His3.3A-PA; His3.3A-PC; His3.3A-PD; histone ; Histone H3 class II; Histone H3 H3.3A H3.3B H3F3 H3F3A H3F3B; Histone H3.3; histone H3.3 ; histone H3.3-like protein; Histone H3.3A; Histone H3.3B; unnamed protein product
Gene Aliases: 9430068D06Rik; CG5825; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; H3-3A; H3-3B; H3-IX; H3-X; H3.3; H3.3A; H3.3B; H3F3; H3F3A; H3F3B; His-3.3A; His3.3; His3.3A; His3.3B; PH3; PP781
UniProt ID: (Bovine) Q5E9F8, (Bovine) A4IFS1, (Human) P84243, (Chicken) P84247, (Mouse) P84244, (Fruit fly) C0HL66
Entrez Gene ID: (Bovine) 326601, (Horse) 100054462, (Bovine) 617051, (Human) 3020, (Chicken) 396233, (Human) 3021, (Mouse) 15078, (Mouse) 15081, (Fruit fly) 33736
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.