Invitrogen
Manufacturer Code:MA531411
Catalog # PIMA531411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL5101 |
Immunogen | Synthetic peptide corresponding to Human HIST1H2BC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: H2B H2BFL H2BFH H2BFG H2BFA H2BFK; H2B histone family, member L; H2B histone family, member S; H2B/a; H2B/g; H2B/h; H2B/k; H2B/l; histone 1, H2bc; histone cluster 1, H2bc; histone H2B; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; Histone H2B.1 A; Histone H2B.a; Histone H2B.g; Histone H2B.h; Histone H2B.k; Histone H2B.l
Gene Aliases: 2610022J01Rik; dJ221C16.3; H2B.1; H2B/l; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFG; H2BFH; H2BFK; H2BFL; H2bfs; HIST1H2BC; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BI; R74621
UniProt ID: (Human) P62807, (Mouse) Q6ZWY9
Entrez Gene ID: (Human) 8347, (Mouse) 68024, (Rat) 680312
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.