Proteintech
Manufacturer Code:158571AP150UL
Catalog # 50-173-0534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Histone H2B Fusion Protein Ag7811 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GL105 H2B H2B.1 H2B/q H2BFQ HIST2H2BE histone cluster 2 H2be Histone H2B Histone H2B GL105 Histone H2B type 2 E Histone H2B.q; H2b 613; H2B histone family, member Q; H2B-clustered histone 21; H2B/q; histone 2, H2be; histone cluster 2, H2be; Histone H2B type 2-E; Histone H2B-GL105; Histone H2B.q
Gene Aliases: AV127319; GL105; H2B; H2b-613; H2B.1; H2BC21; H2BFQ; H2BGL105; H2BQ; HIST2H2BE; RGD1562346; T25626
UniProt ID: (Human) Q16778, (Mouse) Q64524
Entrez Gene ID: (Human) 8349, (Mouse) 319190, (Rat) 295274
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.