Invitrogen
Manufacturer Code:PA141058
Catalog # 01-676-656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 111-125 of human histone H2B &lsqbGenBank no. CAI19747.1] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG17949-PA; H2B; H2B histone family, member F; H2B I; H2B II; H2B III; H2B IV; H2B VI; H2B VII; h2B-143; H2B-clustered histone 3; H2B/f; His2B:CG17949-PA; histone 1, H2bb; Histone H2B; Histone H2B 1/2/3/4/6; Histone H2B 3; Histone H2B type 1-B; Histone H2B.1; Histone H2B.f; RP1-193B12.10 H2B/d H2BFD HIST1H3I MGC125414 MGC125415 MGC125416 MGC9388
Gene Aliases: CG17949; CG33868; CG33870; CG33872; CG33874; CG33876; CG33878; CG33880; CG33882; CG33884; CG33886; CG33888; CG33890; CG33892; CG33894; CG33896; CG33898; CG33900; CG33902; CG33904; CG33906; CG33908; CG33910; Dmel\CG17949; Dmel_CG17949; fj05g08; H2b-143; H2B-I; H2B-II; H2B-III; H2B-IV; H2B-VI; H2B-VII; H2B.1; H2B/f; H2BC3; H2BFF; His2B; His2B:CG17949; His2B:CG33868; His2B:CG33870; His2B:CG33872; His2B:CG33874; His2B:CG33876; His2B:CG33878; His2B:CG33880; His2B:CG33882; His2B:CG33884; His2B:CG33886; His2B:CG33888; His2B:CG33890; His2B:CG33892; His2B:CG33894; His2B:CG33896; His2B:CG33898; His2B:CG33900; His2B:CG33902; His2B:CG33904; His2B:CG33906; His2B:CG33908; His2B:CG33910; HIST1H2BB; hist2h2l; wu:fj05g08; zgc:73093
UniProt ID: (Chicken) P0C1H3, (Human) P33778, (Fruit fly) P02283, (Mouse) Q64475, (Zebrafish) Q6PC60
Entrez Gene ID: (Dog) 608682, (Chicken) 417956, (Human) 3018, (Fruit fly) 326273, (Mouse) 319178, (Zebrafish) 386920
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.