Invitrogen
Manufacturer Code:PA535892
Catalog # PIPA535892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 221 of Histone H1.3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H1 histone family, member 3; H1 VAR.4; H1d; HIST1H1D H1D H1F3 H1.3 histone cluster 1 H1d H1s-2; histone 1, family 3; histone 1, H1d; Histone H1.3; Histone H1c; Histone H1s-2
Gene Aliases: H1-3; H1.3; H1D; H1F3; H1s-2; H1s-4; HIST1H1D
UniProt ID: (Human) P16402, (Mouse) P43277
Entrez Gene ID: (Human) 3007, (Mouse) 14957
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.