Novus Biologicals
Manufacturer Code:NB10056341SS
Catalog # NB10056341T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 194-209 of human HDAC4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HA6116 HD4EC 3.5.1.98 HDAC-A HDACABDMR histone deacetylase 4 histone deacetylase A KIAA0288AHO3; HD4; Histone deacetylase 4; histone deacetylase A
Gene Aliases: AHO3; BDMR; HA6116; HD4; HDAC-4; HDAC-A; HDAC4; HDACA; KIAA0288
UniProt ID: (Human) P56524
Entrez Gene ID: (Human) 9759
Molecular Function: deacetylase hydrolase nucleic acid binding oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.