Novus Biologicals
Manufacturer Code:NBP290171AF532
Catalog # NB037855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 34 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Histamine N-Methyltransferase/HNMT (AAH20677.1 Met1-Ala292). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.8 histamine N-methyltransferase HMT HNMT-S1 HNMT-S2; Histamine N-methyltransferase; HMT
Gene Aliases: HMT; HNMT; HNMT-S1; HNMT-S2; MRT51
UniProt ID: (Human) P50135
Entrez Gene ID: (Human) 3176
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.