Novus Biologicals
Manufacturer Code:NBP16913420UL
Catalog # NBP16913420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to HNMT (histamine N-methyltransferase) The peptide sequence was selected from the N terminal of HNMT. Peptide sequence PGIIGRIGDTKSEIKILSIGGGAGEIDLQILSKVQAQYPGVCINNEVVEP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.8 histamine N-methyltransferase HMT HNMT-S1 HNMT-S2; Histamine N-methyltransferase; HMT
Gene Aliases: HMT; HNMT; HNMT-S1; HNMT-S2; MRT51
UniProt ID: (Human) P50135
Entrez Gene ID: (Human) 3176
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.