Novus Biologicals
Manufacturer Code:NBP303549100UL
Catalog # NB094043
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 194-304 of human Histamine H4R (NP_067637.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AXOR35; AXOR35H4 GPCR105MGC133027 G-protein coupled receptor 105 GPRv53histamine H4 receptor H4Rpfi-013 HH4RBG26 histamine receptor H4 Pfi-013 SP9144; G-protein coupled receptor 105; GPRv53; H4R; Histamine H4 receptor; Pfi-013; SP9144
Gene Aliases: AXOR35; BG26; GPCR105; GPRv53; H4; H4R; HH4R; HRH4
UniProt ID: (Human) Q9H3N8
Entrez Gene ID: (Human) 59340
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.