Novus Biologicals
Manufacturer Code:NBP178790
Catalog # NBP178790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SHNSHKTSLRLNNS corresponding to internal region according to NP_001124527.1 NP_071640.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: gastric receptor 1; gastric receptor 1 Gastric receptor I H2R HH2R histamine H2 receptor histamine receptor H2; Gastric receptor I; H2R; HH2R; Histamine H2 receptor
Gene Aliases: H2R; HRH2
UniProt ID: (Human) P25021
Entrez Gene ID: (Human) 3274
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.