R&D Systems
Manufacturer Code:MAB4726
Catalog # MAB4726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 480054 |
Immunogen | NS0 mouse myeloma cell line transfected with human Histamine H1 R Met1-Ser487 Accession # AAH60802 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: H1R; H1R H1-R hisH1 Histamine H1R histamine receptor H1 HRH1 subclass H1; Histamine H1 receptor; histamine receptor, subclass H1
Gene Aliases: H1-R; H1R; HH1R; hisH1; HRH1
UniProt ID: (Human) P35367
Entrez Gene ID: (Human) 3269
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.