Invitrogen
Manufacturer Code:PA571467
Catalog # PIPA571467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full Length Recombit DEP-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD148; CD148 antigen; Density-enhanced phosphatase 1; DEP-1; HPTP eta; human density enhanced phosphatase-1; protein tyrosine phosphatase, receptor type, J polypeptide; Protein-tyrosine phosphatase eta; Protein-tyrosine phosphatase receptor type J; R-PTP-J; Receptor-type tyrosine-protein phosphatase eta; Receptor-type tyrosine-protein phosphatase eta Protein-tyrosine phosphatase eta R-PTP-eta Density-enhanced phosphatase 1 DEP-1 HPTP eta Protein-tyrosine phosphatase receptor type J R-PTP-J CD148 PTPRJ DEP1; susceptibility to colon cancer 1, mouse, homolog of
Gene Aliases: CD148; DEP1; HPTPeta; PTPRJ; R-PTP-ETA; SCC1
UniProt ID: (Human) Q12913
Entrez Gene ID: (Human) 5795
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.