Invitrogen
Manufacturer Code:MA537953
Catalog # PIMA537953
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2235 |
Immunogen | A synthesized peptide derived from human Hippocalcin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: BDR2 calcium-binding protein calcium-binding protein BDR-2 DYT2 hippocalcin Hpca hypothetical protein MGC73126 neuron-specific calcium-binding protein hippocalcin P23K zgc:73126; calcium-binding protein; Calcium-binding protein BDR-2; Neuron-specific calcium-binding protein hippocalcin; P23K
Gene Aliases: BDR2; DYT2; HPCA
UniProt ID: (Human) P84074, (Mouse) P84075, (Rat) P84076
Entrez Gene ID: (Human) 3208, (Mouse) 15444, (Rat) 29177
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.