Novus Biologicals
Manufacturer Code:NBP254705
Catalog # NBP254705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a Recombinant Protein corresponding to amino acids:MVSSVMKMPEDESTPEKRTEKIFRQMDT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BDR2 Calcium-binding protein BDR-2 hippocalcin neuron specific calcium-binding protein hippocalcin neuron-specific calcium-binding protein hippocalcin; Calcium-binding protein BDR-2; Neuron-specific calcium-binding protein hippocalcin
Gene Aliases: BDR2; DYT2; HPCA
UniProt ID: (Human) P84074
Entrez Gene ID: (Human) 3208
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.