Novus Biologicals
Manufacturer Code:NBP122406R
Catalog # NBP122406R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0090-8L46 |
Immunogen | Mouse recombinant protein of Hip extracellular domain |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: FLJ90230 hedgehog interacting protein hedgehog-interacting protein HIPFLJ20992; Hedgehog-interacting protein; HHIP
Gene Aliases: HHIP; HIP; UNQ5825/PRO19644
UniProt ID: (Human) Q96QV1
Entrez Gene ID: (Human) 64399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.