Invitrogen
Manufacturer Code:PA523484
Catalog # PIPA523484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (76 VPSAFEKQGSAKLEKAEI 93) of Hey1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BHLHb31 CHF2 HERP2 HESR1 HRT-1 OAF1 HES-related repressor protein 1 HES-related repressor protein 2 basic helix-loop-helix protein OAF1 cardiovascular helix-loop-helix factor 2 class B basic helix-loop-helix protein 31 hHRT1 hairy and enhancer of split-related protein 1 hairy-related transcription factor 1 hairy/enhancer-of-split related with YRPW motif protein 1; hairy and enhancer of split-related protein 1; hairy-related transcription factor 1; Hairy/E(spl)-related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif protein 1; HRT-1; mHRT1
Gene Aliases: AI316788; AI414254; bHLHb31; CHF2; Herp2; hesr-1; Hesr1; HRT1
Entrez Gene ID: (Mouse) 15213
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.