Novus Biologicals
Manufacturer Code:NBP237505
Catalog # NBP237505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F10 |
Immunogen | Purified recombinant fragment of human HEXA expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-hexosaminidase subunit alpha; beta-hexosaminidase subunit alpha Beta-N-acetylhexosaminidase subunit alpha EC 3.2.1 EC 3.2.1.52 hexosaminidase A (alpha polypeptide) Hexosaminidase subunit A MGC99608 N-acetyl-beta-glucosaminidase subunit alpha TSD; Beta-N-acetylhexosaminidase subunit alpha; hexosaminidase A (alpha polypeptide); Hexosaminidase subunit A; N-acetyl-beta-glucosaminidase subunit alpha
Gene Aliases: HEXA; TSD
UniProt ID: (Human) P06865
Entrez Gene ID: (Human) 3073
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.