Novus Biologicals
Manufacturer Code:NBP216816
Catalog # NBP216816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human Hexokinase III. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:D-hexose 6-phosphotransferase; ATP:D-hexose 6-phosphotransferase EC 2.7.1 EC 2.7.1.1 hexokinase 3 (white cell) Hexokinase type III hexokinase-3 HK III HKIII HXK3; hexokinase 3 (white cell); Hexokinase type III; Hexokinase-3; Hexokinase-C; HK III
Gene Aliases: HK3; HKIII; HXK3
UniProt ID: (Human) P52790
Entrez Gene ID: (Human) 3101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.