Novus Biologicals
Manufacturer Code:NBP104295R
Catalog # NBP104295R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D7 |
Immunogen | Recombinant human Hexokinase1 (1-917aa) purified from E. coli |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Brain form hexokinase; Brain form hexokinase EC 2.7.1 EC 2.7.1.1 glycolytic enzyme hexokinase 1 Hexokinase type I hexokinase-1 HK I HK1-ta HK1-tb HK1-tc HKI HXK1; glycolytic enzyme; hexokinase I; hexokinase IR; Hexokinase type I; Hexokinase-1; Hexokinase-A; HK I
Gene Aliases: hexokinase; HK; HK1; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1
UniProt ID: (Human) P19367
Entrez Gene ID: (Human) 3098
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.