Invitrogen
Manufacturer Code:MA537951
Catalog # PIMA537951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2232 |
Immunogen | A synthesized peptide derived from human Hex |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: hematopoietically expressed homeobox hematopoietically expressed homeobox related sequence 2 hematopoietically-expressed homeobox protein Hhex HEX Hex1 HHEX Hhex-rs2 HMPH homeobox Homeobox protein HEX homeobox protein PRH homeobox hematopoietically expressed HOX11L-PEN mHex PRH Prhx proline-rich homeodomain-containing transcription factor; Hematopoietically-expressed homeobox protein HHEX; Homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; proline-rich homeodomain-containing transcription factor
Gene Aliases: HEX; HHEX; HMPH; HOX11L-PEN; PRH; PRHX
UniProt ID: (Human) Q03014
Entrez Gene ID: (Human) 3087
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.