Invitrogen
Manufacturer Code:PA519625
Catalog # PIPA519625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 200 to the C-terminus of Human Hex. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hematopoietically expressed homeobox, related sequence 2; Hematopoietically-expressed homeobox protein HHEX; HEX HMPH HOX11L-PEN PRH PRHX hematopoietically-expressed homeobox protein HHEX homeobox protein HEX homeobox protein PRH homeobox hematopoietically expressed proline-rich homeodomain-containing transcription factor; Homeobox protein HEX; Homeobox protein PRH; homeobox, hematopoietically expressed; mHex; proline-rich homeodomain-containing transcription factor
Gene Aliases: HEX; Hex1; HHEX; Hhex-rs2; HMPH; HOX11L-PEN; PRH; PRHX
UniProt ID: (Human) Q03014, (Mouse) Q9CRV1
Entrez Gene ID: (Human) 3087, (Mouse) 15242
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.