Invitrogen
Manufacturer Code:PA5113635
Catalog # PIPA5113635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the c terminal region of mouse Hhex |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hematopoietically expressed homeobox hematopoietically expressed homeobox related sequence 2 Hematopoietically-expressed homeobox protein Hhex HEX Hex1 Hhex Hhex-rs2 HMPH homeobox Homeobox protein HEX Homeobox protein PRH homeobox hematopoietically expressed HOX11L-PEN mHex Prh Prhx proline-rich homeodomain-containing transcription factor; hematopoietically expressed homeobox, related sequence 2; Hematopoietically-expressed homeobox protein Hhex; Homeobox protein HEX; Homeobox protein PRH; mHex
Gene Aliases: Hex; Hex1; Hhex; Hhex-rs2; Prh; Prhx
UniProt ID: (Mouse) Q9CRV1
Entrez Gene ID: (Mouse) 15242
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.