Invitrogen
Manufacturer Code:MA524218
Catalog # PIMA524218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 506220 |
Immunogen | Mouse myeloma cell line NS0-derived recombit human Hepsin Ile163-Leu417 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Serine protease hepsin; Serine protease hepsin catalytic chain; Serine protease hepsin non-catalytic chain; testicular tissue protein Li 85; TMPRSS1 serine protease hepsin serine protease hepsin catalytic chain serine protease hepsin non-catalytic chain transmembrane protease serine 1 transmembrane protease serine 1; Transmembrane protease serine 1
Gene Aliases: HPN; TMPRSS1
UniProt ID: (Human) P05981
Entrez Gene ID: (Human) 3249
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.