R&D Systems
Manufacturer Code:AF4776SP
Catalog # AF4776SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Hepsin Ser46-Leu417 with Asp161Glu and Arg162Lys substitutions Accession # P05981 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.21 EC 3.4.21.106 hepsin HPN serine protease hepsin serine protease hepsin catalytic chain TMPRSS1 TMPRSS1serine protease hepsin non-catalytic chain Transmembrane protease serine 1 transmembrane protease serine 1; Serine protease hepsin; Serine protease hepsin catalytic chain; Serine protease hepsin non-catalytic chain; testicular tissue protein Li 85; Transmembrane protease serine 1
Gene Aliases: HPN; TMPRSS1
UniProt ID: (Human) P05981
Entrez Gene ID: (Human) 3249
Molecular Function:
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.