Invitrogen
Manufacturer Code:PA577256
Catalog # PIPA577256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 41-90 of Human Heparin Cofactor II. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HC-II; Heparin cofactor 2; Heparin cofactor 2 Heparin cofactor II HC-II Protease inhibitor leuserpin-2 HLS2 Serpin D1 SERPIND1 HCF2; Heparin cofactor II; HLS2; leuserpin 2; leuserpin-2; Protease inhibitor leuserpin-2; serine (or cysteine) peptidase inhibitor, clade D, member 1; serine (or cysteine) proteinase inhibitor, clade D (heparin cofactor), member 1; serine (or cysteine) proteinase inhibitor, clade D, member 1; Serpin D1; serpin peptidase inhibitor, clade D (heparin cofactor), member 1
Gene Aliases: AA985900; AI303446; D22S673; HC2; HCF2; HCII; HLS2; LS2; rls2var1; SERPIND1; THPH10
UniProt ID: (Human) P05546, (Rat) Q64268, (Mouse) P49182
Entrez Gene ID: (Human) 3053, (Rat) 79224, (Mouse) 15160
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.