Invitrogen
Manufacturer Code:PA547108
Catalog # PIPA547108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Serpin D1/Heparin Cofactor II Glu24-Ser478 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: D22S673 HC2 HCF2 HCII HLS2 LS2 heparin cofactor 2 leuserpin 2 protease inhibitor leuserpin-2 serine (or cysteine) proteinase inhibitor clade D (heparin cofactor) member 1 serpin D1; HC-II; Heparin cofactor 2; Heparin cofactor II; Protease inhibitor leuserpin-2; serine (or cysteine) proteinase inhibitor, clade D (heparin cofactor), member 1; Serpin D1
Gene Aliases: AA985900; AI303446; Hcf2; HCII; Serpind1
UniProt ID: (Mouse) P49182
Entrez Gene ID: (Mouse) 15160
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.