Novus Biologicals
Manufacturer Code:NBP2840640.1ML
Catalog # NB031992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of human Heparanase/HPSE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.2 endo-glucoronidase HEP heparanase Heparanase-1 Hpa1 HPAheparanase exon 9 and 10 deletion HPR1 HPSE1heparanase-1 HSE1heparanase exon10-deletion; Endo-glucoronidase; Heparanase; Heparanase 50 kDa subunit; Heparanase 8 kDa subunit; Heparanase-1; Hpa1
Gene Aliases: HEP; HPA; HPA1; HPR1; HPSE; HPSE1; HSE1
UniProt ID: (Human) Q9Y251
Entrez Gene ID: (Human) 10855
Molecular Function: glycosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.