R&D Systems
Manufacturer Code:AF7276SP
Catalog # AF7276SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived human Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3 Gly60-Asp390 Accession # Q9Y662 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-3B; 3OST3B1 h3-OST-3B heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1 Heparan Sulfate Glucosamine 3OSulfotransferase 3; heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1; heparan sulfate 3-O-sulfotransferase 3B1; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3B1; Heparan sulfate glucosamine 3-O-sulfotransferase 3B1
Gene Aliases: 3-OST-3B; 3OST3B1; h3-OST-3B; HS3ST3B; HS3ST3B1
UniProt ID: (Human) Q9Y662
Entrez Gene ID: (Human) 9953
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.