Novus Biologicals
Manufacturer Code:NBP287562
Catalog # NB035184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp761K2315 heparan sulfate 6-O-sulfotransferase 3; Heparan-sulfate 6-O-sulfotransferase 3; HS6ST-3
Gene Aliases: HS6ST-3; HS6ST3
UniProt ID: (Human) Q8IZP7
Entrez Gene ID: (Human) 266722
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.