R&D Systems
Manufacturer Code:AF5057SP
Catalog # AF5057SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO cell-derived recombinant human HS6ST1 Pro28-Trp401 Accession # AAY14736 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp547H098 EC 2.8.2 EC 2.8.2.- FLJ25392 heparan sulfate 6-O-sulfotransferase 1 heparan-sulfate 6-sulfotransferase HS6ST-1 HS6STheparan-sulfate 6-O-sulfotransferase 1 MGC116899 MGC116901; Heparan-sulfate 6-O-sulfotransferase 1; heparan-sulfate 6-sulfotransferase; HS6ST-1
Gene Aliases: HH15; HS6ST; HS6ST1
UniProt ID: (Human) O60243
Entrez Gene ID: (Human) 9394
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.