R&D Systems
Manufacturer Code:MAB60851SP
Catalog # MAB60851SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 712011 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Gly184-Lys456 Accession # Q9Y661 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-OST-4; 30ST4 3-OST-4 3OST4 heparan sulfate 3-O-sulfotransferase-4 EC 2.8.2 EC 2.8.2.23 h3-OST-4 heparan sulfate (glucosamine) 3-O-sulfotransferase 4 Heparan sulfate 3-O-sulfotransferase 4 Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4 heparan sulfate glucosamine 3-O-sulfotransferase 4; heparan sulfate (glucosamine) 3-O-sulfotransferase 4; heparan sulfate 3-O-sulfotransferase 4; heparan sulfate 3-O-sulfotransferase-4; Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4; Heparan sulfate glucosamine 3-O-sulfotransferase 4
Gene Aliases: 3-OST-4; 30ST4; 3OST4; h3-OST-4; HS3ST4
UniProt ID: (Human) Q9Y661
Entrez Gene ID: (Human) 9951
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.