Invitrogen
Manufacturer Code:MA535760
Catalog # PIMA535760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1444 |
Immunogen | A synthesized peptide derived from human Hemopexin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-1B-glycoprotein; Beta-1B-glycoprotein Hemopexin HPX Hpxn HX Hyaluronidase; Hemopexin
Gene Aliases: HPX; Hpxn; HX
UniProt ID: (Human) P02790, (Mouse) Q91X72
Entrez Gene ID: (Human) 3263, (Mouse) 15458
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.