Invitrogen
Manufacturer Code:ABS0130402
Catalog # ABA-130402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | Hemopexin purified from human plasma. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-1B-glycoprotein; Hemopexin
Gene Aliases: HPX; HX
UniProt ID: (Human) P02790
Entrez Gene ID: (Human) 3263
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.