Invitrogen
Manufacturer Code:MA535794
Catalog # PIMA535794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1510 |
Immunogen | A synthesized peptide derived from human Hemoglobin subunit alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1/2-globin; Alpha-1/2-globin Alpha-globin GloA Hba Hba1 HBA2 Hba-a1 Hba-a2 HBAM Hbat1 HBA-T3 HBH Hemoglobin alpha chain Hemoglobin alpha-1/2 chain Hemoglobin subunit alpha Hemoglobin subunit alpha-1/2; alpha-globin; globin, alpha; hemoglobin alpha 1 chain; hemoglobin alpha, adult chain 1; hemoglobin alpha, adult chain 2; Hemoglobin alpha-1/2 chain; Hemoglobin subunit alpha-1/2
Gene Aliases: GloA; Hba-a1; Hba-a2; Hba1; HBAM
UniProt ID: (Rat) P01946
Entrez Gene ID: (Rat) 25632, (Rat) 287167
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.