Invitrogen
Manufacturer Code:PA19527
Catalog # 01-674-410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide mixture based on the human hemoglobin alpha protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha one globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin, alpha 1
Gene Aliases: HBA-T3; HBA1; HBA2; HBH
UniProt ID: (Human) P69905
Entrez Gene ID: (Human) 3039
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.