R&D Systems
Manufacturer Code:MAB3915SP
Catalog # MAB3915SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 394903 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Hck Met22-Asp80 Accession # P08631 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Bmk EC 2.7.10 EC 2.7.10.2 Hctk hemopoietic cell kinaseJTK9 JTK9 p59-HCK/p60-HCK tyrosine-protein kinase HCK; Hematopoietic cell kinase; Hemopoietic cell kinase; p59-HCK/p60-HCK; p59Hck; p61Hck; Tyrosine-protein kinase HCK
Gene Aliases: HCK; JTK9; p59Hck; p61Hck
UniProt ID: (Human) P08631
Entrez Gene ID: (Human) 3055
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.