Proteintech
Manufacturer Code:113581AP150UL
Catalog # 50-172-7090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Harmonin Fusion Protein Ag1919 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIE 75 AIE75 Antigen NY CO 38/NY CO 37 Harmonin NY CO 37 NY CO 38 PDZ 45 PDZ 73 PDZ 73/NY CO 38 PDZ73 USH1C Usher 1C; Antigen NY-CO-38/NY-CO-37; Autoimmune enteropathy-related antigen AIE-75; Harmonin; harmonin a1; PDZ domain-containing protein; Protein PDZ-73; Renal carcinoma antigen NY-REN-3; Usher syndrome 1C (autosomal recessive, severe); Usher syndrome 1C homolog; Usher syndrome type-1C protein; Usher syndrome type-1C protein homolog
Gene Aliases: 2010016F01Rik; AIE-75; AIE75; DFNB18; DFNB18A; harmonin; NY-CO-37; NY-CO-38; PDZ-45; PDZ-73; PDZ-73/NY-CO-38; PDZ73; PDZD7C; USH1C; ush1cpst
UniProt ID: (Human) Q9Y6N9, (Mouse) Q9ES64
Entrez Gene ID: (Human) 10083, (Rat) 308596, (Mouse) 72088
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.