Invitrogen
Manufacturer Code:PA569136
Catalog # PIPA569136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human Hao2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (S)-2-hydroxy-acid oxidase, peroxisomal; (S)-hydroxy-acid oxidase, peroxisomal; Cell growth-inhibiting gene 16 protein; glycolate oxidase; Hao2 Hao3 Haox2; HAOX2; hydroxyacid oxidase (glycolate oxidase) 3; Hydroxyacid oxidase 2; hydroxyacid oxidase 2 (long chain); hydroxyacid oxidase 2, kidney; hydroxyacid oxidase 3 (medium-chain); Long chain alpha-hydroxy acid oxidase; Long-chain L-2-hydroxy acid oxidase; Medium chain alpha-hydroxy acid oxidase; medium-chain 2-hydroxy acid oxidase HAOX3; Medium-chain L-2-hydroxy acid oxidase
Gene Aliases: AI325478; GIG16; Hao-2; HAO2; Hao3; HAOX2; Haox3
UniProt ID: (Human) Q9NYQ3, (Bovine) Q3ZBW2, (Rat) Q07523, (Mouse) Q9NYQ2
Entrez Gene ID: (Human) 51179, (Pig) 100522133, (Bovine) 509481, (Dog) 475814, (Horse) 100066884, (Rat) 84029, (Mouse) 56185
Molecular Function:
dehydrogenase
isomerase
ligase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.