Invitrogen
Manufacturer Code:MA146047
Catalog # PIMA146047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1/24 |
Immunogen | Hairy gene product. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: barrel; CG6494-PA; CG6494-PB; Dmel&lowbarCG6494 8247 CG6494 DmelCG6494 Hairy bHLHb39 bHLHc14 brr dDr1 l(3)08247 l(3)rM384; h-PA; h-PB; Protein hairy; RE40955p
Gene Aliases: 8247; bHLHb39; bHLHc14; brr; CG6494; dDr1; Dmel\CG6494; Dmel_CG6494; H; Hairy; HRY; l(3)08247; l(3)rM384
UniProt ID: (Fruit fly) P14003
Entrez Gene ID: (Fruit fly) 38995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.