Novus Biologicals
Manufacturer Code:NBP271850FR
Catalog # NBP271850FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E1 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1787-2144 of human HYPB (NP_054878) produced in E.coli. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ22472 FLJ23184 FLJ45883 FLJ46217 HIF1 HIF-1EC 2.1.1.43 HIP-1 histone-lysine N-methyltransferase SETD2 hSET2 huntingtin interacting protein 1 Huntingtin yeast partner B Huntingtin-interacting protein 1 Huntingtin-interacting protein B HYPBp231HBP KIAA1732SET domain-containing protein 2 KMT3AHBP231 Lysine N-methyltransferase 3A SET domain containing 2 SET2FLJ16420; HIF-1; HIP-1; Histone-lysine N-methyltransferase SETD2; hSET2; huntingtin interacting protein 1; Huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; Lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain-containing protein 2
Gene Aliases: HBP231; HIF-1; HIF1; HIP-1; HSPC069; HYPB; KIAA1732; KMT3A; LLS; p231HBP; SET2; SETD2
UniProt ID: (Human) Q9BYW2
Entrez Gene ID: (Human) 29072
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.