Proteintech
Manufacturer Code:181391AP150UL
Catalog # 50-173-2009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HYAL4 Fusion Protein Ag12766 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chondroitin sulfate endo-beta-N-acetylgalactosaminidase; Chondroitin sulfate hydrolase; CSHY; Hyal 4 HYAL4 Hyaluronidase 4 hyaluronoglucosaminidase 4; Hyal-4; hyaluronidase 4; Hyaluronidase-4; Hyaluronoglucosaminidase-4
Gene Aliases: 4632428M18Rik; CSHY; HYAL4
UniProt ID: (Human) Q2M3T9, (Mouse) Q05A56
Entrez Gene ID: (Human) 23553, (Rat) 404783, (Mouse) 77042
Molecular Function: glycosidase hydrolase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.