Invitrogen
Manufacturer Code:PA579422
Catalog # PIPA579422
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human HYAL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hyal-3; hyaluronidase 3; Hyaluronidase-3; Hyaluronidase-3 Hyal-3; Hyaluronoglucosaminidase-3; LuCa-3; Lung carcinoma protein 3
Gene Aliases: HYAL-3; HYAL3; Hyl3; LUCA-3; LUCA3
UniProt ID: (Human) O43820, (Mouse) Q8VEI3
Entrez Gene ID: (Human) 8372, (Mouse) 109685
Molecular Function:
glycosidase
hydrolase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.