Invitrogen
Manufacturer Code:PA5114182
Catalog # PIPA5114182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: RAAMACSHQR CHGHGRCARR DPGQMEAFLH LWPDGSLGDW KSFSCHCYWG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hyal-3; HYAL3 Hyal-3 hyaluronidase 3 Hyaluronidase-3 hyaluronoglucosaminidase 3 Hyaluronoglucosaminidase-3 Hyl3 LUCA14 LUCA3 LuCa-3 Lung carcinoma protein 3 Minna14; Hyaluronidase-3; Hyaluronoglucosaminidase-3; LuCa-3; Lung carcinoma protein 3
Gene Aliases: HYAL-3; HYAL3; LUCA-3; LUCA3
UniProt ID: (Human) O43820
Entrez Gene ID: (Human) 8372
Molecular Function: glycosidase hydrolase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.