Invitrogen
Manufacturer Code:PA5115387
Catalog # PIPA5115387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HYAL2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI256841 AU020858 AW555733 HYAL2 Hyal-2 hyaluronidase 2 hyaluronidase-2 hyaluronoglucosaminidase 2 hyaluronoglucosaminidase-2 LUCA2 LuCa-2 Lung carcinoma protein 2 lysosomal hyaluronidase PH20 homolog PH-20 homolog; Hyal-2; hyaluronidase 2; Hyaluronidase-2; Hyaluronoglucosaminidase-2; LuCa-2; Lung carcinoma protein 2; lysosomal hyaluronidase; PH-20 homolog; PH20 homolog
Gene Aliases: AI256841; AU020858; AW555733; HYAL2; LUCA2
UniProt ID: (Human) Q12891, (Mouse) O35632
Entrez Gene ID: (Human) 8692, (Rat) 64468, (Mouse) 15587
Molecular Function:
glycosidase
hydrolase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.