Novus Biologicals
Manufacturer Code:NBP181283
Catalog # NBP181283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:APPIFTGRPFVVAWDVPTQDCGPRLKVPLDLNAFDVQASPNEGFVNQNITIFYRDRLGLYPRFDSAGRSVHGGVPQNVSLWAHRK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.2.1.35 Hyal-2 hyaluronidase 2 hyaluronoglucosaminidase 2 hyaluronoglucosaminidase-2 LuCa-2 LUCA2hyaluronidase-2 Lung carcinoma protein 2 lysosomal hyaluronidase PH20 homolog PH-20 homolog; Hyal-2; hyaluronidase 2; Hyaluronidase-2; Hyaluronoglucosaminidase-2; LuCa-2; Lung carcinoma protein 2; lysosomal hyaluronidase; PH-20 homolog; PH20 homolog
Gene Aliases: HYAL2; LUCA2
UniProt ID: (Human) Q12891
Entrez Gene ID: (Human) 8692
Molecular Function:
glycosidase
hydrolase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.