Novus Biologicals
Manufacturer Code:NBP30499520UL
Catalog # NB094569
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 367-473 of human HYAL2 (NP_149348.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.2.1.35 Hyal-2 hyaluronidase 2 hyaluronoglucosaminidase 2 hyaluronoglucosaminidase-2 LuCa-2 LUCA2hyaluronidase-2 Lung carcinoma protein 2 lysosomal hyaluronidase PH20 homolog PH-20 homolog; Hyal-2; hyaluronidase 2; Hyaluronidase-2; Hyaluronoglucosaminidase-2; LuCa-2; Lung carcinoma protein 2; lysosomal hyaluronidase; PH-20 homolog; PH20 homolog
Gene Aliases: HYAL2; LUCA2
UniProt ID: (Human) Q12891
Entrez Gene ID: (Human) 8692
Molecular Function: glycosidase hydrolase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.