Invitrogen
Manufacturer Code:PA579421
Catalog # PIPA579421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of mouse HYAL1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hyal-1; hyaluronidase 1; Hyaluronidase-1; Hyaluronidase-1 Hyal-1 Hyaluronoglucosaminidase-1 Hyal1; Hyaluronoglucosaminidase-1
Gene Aliases: Hya1; Hyal-1; Hyal1
UniProt ID: (Mouse) Q91ZJ9
Entrez Gene ID: (Mouse) 15586
Molecular Function: glycosidase hydrolase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.