Invitrogen
Manufacturer Code:PA527901
Catalog # PIPA527901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 3 and 374 of Human HYAL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hyal-1; HYAL-1 LUCA1 NAT6 hyaluronidase 1 hyaluronidase-1 hyaluronoglucosaminidase-1 luCa-1 lung carcinoma protein 1 plasma hyaluronidase tumor suppressor LUCA-1; Hyaluronidase-1; Hyaluronoglucosaminidase-1; LuCa-1; Lung carcinoma protein 1; plasma hyaluronidase; tumor suppressor LUCA-1
Gene Aliases: HYAL-1; HYAL1; LUCA1; MPS9; NAT6
UniProt ID: (Human) Q12794
Entrez Gene ID: (Human) 3373
Molecular Function:
glycosidase
hydrolase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.