Proteintech
Manufacturer Code:251791AP150UL
Catalog # 50-173-4772
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HYAL1 Fusion Protein Ag17943 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HYAL 1 HYAL1 Hyaluronidase 1 hyaluronoglucosaminidase 1 LuCa 1 LUCA1 Lung carcinoma protein 1 NAT6; Hyal-1; hyaluronidase 1; Hyaluronidase-1; Hyaluronoglucosaminidase-1; LuCa-1; Lung carcinoma protein 1; plasma hyaluronidase; tumor suppressor LUCA-1
Gene Aliases: Hya1; HYAL-1; HYAL1; LUCA1; MPS9; NAT6
UniProt ID: (Human) Q12794, (Mouse) Q91ZJ9
Entrez Gene ID: (Human) 3373, (Mouse) 15586
Molecular Function:
glycosidase
hydrolase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.